siRNA Details
Gene Symbol: Elac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053255.3 | NP_444485.2 | 4 | 2057 | ||
GenBank mRNA | AK018424.1 | - | 1 | 1638 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
