siRNA Details
Gene Symbol: Spdyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029048.3 | NP_083324.3 | 2 | 352 | ||
GenBank mRNA | AK015385.1 | - | 2 | 331 | ||
AK015441.1 | - | 2 | 353 | |||
AY820308.1 | - | 2 | 277 | |||
BC141200.1 | - | 2 | 331 | 9055812 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
