siRNA Details
Gene Symbol: Pcf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017569.1 | - | 1 | 999 | ||
BC017150.1 | - | 1 | 978 | |||
BC030309.1 | - | 1 | 1033 | |||
BC037632.1 | - | 1 | 1017 | |||
BC048838.1 | - | 1 | 1001 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
