siRNA Details
Gene Symbol: Iqca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029122.2 | NP_083398.2 | 2 | 395 | ||
GenBank mRNA | AK015512.1 | - | 2 | 396 | ||
AK016353.1 | - | 2 | 396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
