siRNA Details
Gene Symbol: Sf3a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029157.3 | NP_083433.1 | 4 | 249 | ||
GenBank mRNA | AK015776.1 | - | 4 | 228 | ||
AK030743.1 | - | 4 | 284 | |||
AK150733.1 | - | 4 | 247 | |||
AK151141.1 | - | 4 | 247 | |||
AK151382.1 | - | 4 | 247 | |||
AK161646.1 | - | 4 | 270 | |||
AK168966.1 | - | 4 | 273 | |||
AK169324.1 | - | 4 | 293 | |||
BC009141.1 | - | 4 | 249 | 3598647 | ||
BC092058.1 | - | 4 | 281 | 4017388 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
