siRNA Details
Gene Symbol: Gm10413 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029288.3 | NP_083564.2 | 3 | 467 | ||
GenBank mRNA | AK005583.1 | - | 3 | 457 | ||
BC100412.1 | - | 3 | 467 | 30916569 |
Gene Symbol: Gm5458 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024706.2 | NP_001019877.2 | 3 | 473 | ||
GenBank mRNA | BC060995.1 | - | 3 | 473 | 6774503 |
Gene Symbol: Gm5795 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270806.1 | NP_001257735.1 | 3 | 438 | ||
GenBank mRNA | BU946301.1 | - | 3 | 441 |
Gene Symbol: Gm5797 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025085.2 | NP_001020256.1 | 3 | 467 | ||
GenBank mRNA | BC090991.1 | - | 3 | 467 | 6742820 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
