siRNA Details
Gene Symbol: Pibf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029320.3 | NP_083596.2 | 18 | 3118 | ||
GenBank mRNA | AK006060.1 | - | 9 | 1092 | ||
AL361753.1 | - | 3 | 212 | |||
BC038269.1 | - | 5 | 724 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
