siRNA Details
Gene Symbol: 4933411G11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177880.4 | NP_808548.1 | 8 | 1472 | ||
GenBank mRNA | AK030175.1 | - | 8 | 1473 | ||
BC115568.1 | - | 8 | 1005 | 40102090 | ||
BC115569.1 | - | 8 | 1094 | 40102091 |
Gene Symbol: Spdyb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029048.3 | NP_083324.3 | 7 | 1155 | ||
GenBank mRNA | AK015385.1 | - | 6 | 965 | ||
AK015441.1 | - | 7 | 1156 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
