siRNA Details
Gene Symbol: Pcf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017569.1 | - | 1 | 1567 | ||
BC017150.1 | - | 1 | 1546 | |||
BC030309.1 | - | 1 | 1600 | |||
BC037632.1 | - | 1 | 1585 | |||
BC048838.1 | - | 1 | 1569 |

More Information
Other Species Targeted

