siRNA Details
Gene Symbol: Sf3a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029157.3 | NP_083433.1 | 12 | 989 | ||
GenBank mRNA | AK030743.1 | - | 12 | 1024 | ||
AK150733.1 | - | 12 | 987 | |||
AK151141.1 | - | 12 | 987 | |||
AK151382.1 | - | 12 | 987 | |||
AK161646.1 | - | 12 | 1010 | |||
AK168966.1 | - | 12 | 1013 | |||
AK169324.1 | - | 12 | 1033 | |||
AK187073.1 | - | 5 | 389 | |||
AK213498.1 | - | 2 | 131 | |||
AK215652.1 | - | 2 | 131 | |||
BC009141.1 | - | 12 | 989 | 3598647 | ||
BC092058.1 | - | 12 | 1021 | 4017388 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
