siRNA Details
Gene Symbol: Sall4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017633.1 | - | 1 | 806 | ||
AK145388.1 | - | 4 | 4185 | |||
BC067396.1 | - | 1 | 608 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
