siRNA Details
Gene Symbol: 9130401M01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029418.4 | NP_083694.1 | 2, 3 | 306 | ||
GenBank mRNA | AK018655.1 | - | 2, 3 | 230 | ||
AK047927.1 | - | 2, 3 | 273 | |||
AK089715.1 | - | 230 | ||||
AK167012.1 | - | 2, 3 | 328 | |||
AK185510.1 | - | 1, 2 | 100 | |||
AY053393.1 | - | 2, 3 | 329 | |||
BC050895.1 | - | 2, 3 | 306 | 5714794 |
