siRNA Details
Gene Symbol: Smurf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038627.1 | NP_001033716.1 | 7, 8 | 998 | ||
NM_029438.3 | NP_083714.3 | 7, 8 | 998 | |||
GenBank mRNA | AK015264.1 | - | 3, 4 | 397 | ||
AK154491.1 | - | 7, 8 | 999 | |||
BC029097.1 | - | 7, 8 | 1026 | 3660965 | ||
CB524571.1 | - | 1, 2 | 76 | |||
CX568994.1 | - | 5, 6 | 598 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
