siRNA Details
Gene Symbol: Keg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029550.4 | NP_083826.1 | 2, 3 | 150 | ||
GenBank mRNA | AB028071.1 | - | 2, 3 | 127 | ||
AK002360.1 | - | 2, 3 | 148 | |||
AK165349.1 | - | 2, 3 | 148 | |||
AK165515.1 | - | 2, 3 | 148 | |||
BC010803.1 | - | 2, 3 | 140 | 4238654 |
Gene Symbol: Gm4952 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013762.2 | NP_001013784.2 | 2, 3 | 272 | ||
NM_001167907.1 | NP_001161379.1 | 2, 3 | 210 | |||
GenBank mRNA | BC089619.1 | - | 2, 3 | 200 | 6768245 |

More Information
Other Species Targeted

Gene Ontology Categories:
