siRNA Details
Gene Symbol: Fam65a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081241.2 | NP_001074710.2 | 22 | 3736 | ||
GenBank mRNA | AK010063.1 | - | 4 | 321 | ||
AK052316.1 | - | 23 | 3617 | |||
AK122573.1 | - | 22 | 3710 | |||
BC006820.1 | - | 5 | 522 | |||
BC030451.1 | - | 12 | 2799 | 5400266 | ||
BC079880.1 | - | 22 | 3650 | 6852251 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
