siRNA Details
Gene Symbol: Gbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194336.2 | NP_919317.2 | 11 | 1759 | ||
GenBank mRNA | AK128993.1 | - | 1755 | |||
AK129021.1 | - | 1031 | ||||
AK171913.1 | - | 8 | 3397 | |||
BC007143.1 | - | 10 | 1616 | 3495728 | ||
BC052867.1 | - | 6 | 987 | |||
BC057969.1 | - | 1745 | 4978977 | |||
BC115768.1 | - | 1755 | 40085916 |
Gene Symbol: Gbp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039646.2 | NP_001034735.2 | 11 | 1765 | ||
GenBank mRNA | BC112328.1 | - | 11 | 1782 | 5348193 | |
BC114209.1 | - | 11 | 1765 | 4980612 | ||
DQ985743.1 | - | 11 | 1765 |
Gene Symbol: Gbp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029509.4 | NP_083785.3 | 1518 | |||
GenBank mRNA | AK017988.1 | - | 5 | 735 | ||
BC031475.1 | - | 2435 | 2654170 | |||
BC080665.1 | - | 6 | 958 | |||
DQ295175.1 | - | 9 | 1433 |

More Information
Other Species Targeted

Gene Ontology Categories:
