siRNA Details
Gene Symbol: ATR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184.3 | NP_001175.2 | 21, 22 | 4075 | ||
XM_011512924.1 | XP_011511226.1 | 21, 22 | 4042 | |||
XM_011512925.1 | XP_011511227.1 | 20, 21 | 3850 | |||
XM_011512926.2 | XP_011511228.1 | 21, 22 | 4042 | |||
XM_017006643.1 | XP_016862132.1 | 20, 21 | 3850 | |||
GenBank mRNA | U49844.1 | - | 21, 22 | 4058 | ||
U76308.1 | - | 21, 22 | 4032 | |||
Y09077.1 | - | 21, 22 | 4032 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
