siRNA Details
Gene Symbol: Gm5941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034103.1 | NP_001029275.1 | 477 | |||
GenBank mRNA | BC051629.1 | - | 422 | |||
BC100438.1 | - | 477 | 30916603 |
Gene Symbol: 1700010D01Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005817.1 | - | 2 | 421 | ||
BC059715.1 | - | 2 | 424 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
