siRNA Details
Gene Symbol: Gm14325 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024849.2 | NP_001020020.2 | 4 | 599 | ||
GenBank mRNA | BC092391.1 | - | 3 | 1771 | 6853907 | |
BC092392.1 | - | 4 | 580 |
Gene Symbol: Gm14326 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190302.2 | NP_001177231.1 | 5 | 653 | ||
GenBank mRNA | CV555523.1 | - | 3 | 423 |
Gene Symbol: Gm14393 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085546.2 | NP_001079015.1 | 4 | 419 | ||
GenBank mRNA | CF161713.1 | - | 4 | 419 | ||
CF164689.1 | - | 4 | 413 |
Gene Symbol: Gm14420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177568.1 | NP_001171039.1 | 4 | 624 | ||
GenBank mRNA | BC124559.1 | - | 4 | 624 | 30730885 |
Gene Symbol: Gm6182 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008984.1 | - | 6 | 620 | ||
AK166086.1 | - | 5 | 523 | |||
BC048160.1 | - | 5 | 486 | |||
BC051922.1 | - | 5 | 486 | |||
BC059210.1 | - | 4 | 1620 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
