siRNA Details
Gene Symbol: Ppp1r12c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029834.3 | NP_084110.2 | 1, 2 | 423 | ||
GenBank mRNA | AK010836.1 | - | 1, 2 | 409 | ||
AK046570.1 | - | 1, 2 | 393 | |||
AK144521.1 | - | 1, 2 | 367 | |||
AK144691.1 | - | 1, 2 | 424 | |||
AK187068.1 | - | 1, 2 | 34 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
