siRNA Details
Gene Symbol: 9530003J23Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029906.3 | NP_084182.2 | 1 | 175 | ||
GenBank mRNA | AK020538.1 | - | 1 | 101 | ||
AK035427.1 | - | 1 | 101 | |||
AK035441.1 | - | 1 | 149 | |||
BC120771.1 | - | 1 | 96 | 40129694 | ||
BC120797.1 | - | 1 | 96 | 40129720 | ||
DV040577.1 | - | 216 |
