siRNA Details
Gene Symbol: Tmem116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161626.1 | NP_001155098.1 | 6, 7 | 475 | ||
NM_001161627.1 | NP_001155099.1 | 7, 8 | 580 | |||
NM_029912.2 | NP_084188.1 | 6, 7 | 513 | |||
GenBank mRNA | AK015791.1 | - | 538 | |||
AK021099.1 | - | 6, 7 | 519 | |||
AK039773.1 | - | 1024 | ||||
BC019151.1 | - | 6, 7 | 470 | 5064772 | ||
CK127968.1 | - | 7, 8 | 580 | |||
DV658154.1 | - | 4, 5 | 314 |
