siRNA Details
Gene Symbol: Vps33a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029929.3 | NP_084205.3 | 1, 2 | 144 | ||
GenBank mRNA | AF439858.1 | - | 1, 2 | 128 | ||
AK019463.1 | - | 1, 2 | 128 | |||
AK041982.1 | - | 1, 2 | 140 | |||
AK140553.1 | - | 1, 2 | 133 | |||
AK150282.1 | - | 1, 2 | 136 | |||
AK150550.1 | - | 1, 2 | 136 | |||
AK194964.1 | - | 1, 2 | 58 | |||
BB852750.1 | - | 1, 2 | 168 | |||
BC046417.1 | - | 1, 2 | 135 | 4501207 |
