siRNA Details
Gene Symbol: Fam204a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029648.6 | NP_083924.2 | 2 | 668 | ||
GenBank mRNA | AK075914.1 | - | 2 | 669 | ||
AK162079.1 | - | 2 | 620 | |||
BC048505.1 | - | 3 | 295 | |||
BC061459.1 | - | 467 | 6829614 |
