siRNA Details
Gene Symbol: Gsdma2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029727.2 | NP_084003.2 | 8 | 923 | ||
GenBank mRNA | AK008613.1 | - | 6 | 576 | ||
AK008663.1 | - | 8 | 919 | |||
AK019065.1 | - | 1 | 86 | |||
BC109335.1 | - | 7 | 826 | 40052139 | ||
BC109336.1 | - | 8 | 857 | 40052146 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
