siRNA Details
Gene Symbol: Tbc1d9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290759.1 | NP_001277688.1 | 20 | 3025 | ||
NM_001290760.1 | NP_001277689.1 | 20 | 3028 | |||
NM_029745.2 | NP_084021.2 | 19 | 2977 | |||
GenBank mRNA | AK031992.1 | - | 5 | 447 | ||
AK035457.1 | - | 19 | 2942 | |||
AK088860.1 | - | 14 | 1927 | |||
AK147964.1 | - | 19 | 2942 | |||
AK172999.1 | - | 21 | 2965 | |||
AK184661.1 | - | 3 | 118 | |||
AK186352.1 | - | 2 | 68 | |||
AK188575.1 | - | 2 | 67 | |||
AK192140.1 | - | 2 | 68 | |||
AK194602.1 | - | 3 | 118 | |||
AK195900.1 | - | 3 | 118 | |||
AK197376.1 | - | 44 | ||||
AK199606.1 | - | 3 | 119 | |||
AK207874.1 | - | 2 | 67 | |||
AK207903.1 | - | 2 | 67 | |||
AK210511.1 | - | 2 | 67 | |||
AK212277.1 | - | 3 | 118 | |||
AK214532.1 | - | 2 | 67 | |||
BC048085.1 | - | 10 | 1291 | |||
BC058596.1 | - | 7 | 562 | |||
BC062928.1 | - | 19 | 2964 | 6854669 | ||
BC065080.1 | - | 19 | 2956 | 6826791 | ||
BC065130.1 | - | 19 | 2920 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
