siRNA Details
Gene Symbol: Tbc1d9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290759.1 | NP_001277688.1 | 22 | 3807 | ||
NM_001290760.1 | NP_001277689.1 | 22 | 3810 | |||
NM_029745.2 | NP_084021.2 | 21 | 3759 | |||
GenBank mRNA | AK012224.1 | - | 1 | 629 | ||
AK031992.1 | - | 7 | 1229 | |||
AK088860.1 | - | 16 | 2709 | |||
AK147964.1 | - | 21 | 3724 | |||
AK172999.1 | - | 23 | 3747 | |||
AK183366.1 | - | 1 | 130 | |||
AK190655.1 | - | 1 | 130 | |||
AK191880.1 | - | 1 | 130 | |||
AK197752.1 | - | 1 | 130 | |||
AK212412.1 | - | 1 | 130 | |||
AK214088.1 | - | 1 | 130 | |||
AK219911.1 | - | 1 | 120 | |||
BC048085.1 | - | 12 | 2073 | |||
BC058596.1 | - | 9 | 1344 | |||
BC062928.1 | - | 21 | 3746 | 6854669 | ||
BC065080.1 | - | 21 | 3738 | 6826791 | ||
BC065130.1 | - | 21 | 3702 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
