siRNA Details
Gene Symbol: DNAAF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256714.1 | NP_001243643.1 | 3, 4 | 507 | ||
NM_001256715.1 | NP_001243644.1 | 3, 4 | 433 | |||
NM_178837.4 | NP_849159.2 | 3, 4 | 444 | |||
GenBank mRNA | AK093458.1 | - | 3, 4 | 435 | ||
AK097388.1 | - | 3, 4 | 355 | |||
BC016843.1 | - | 3, 4 | 263 | 3904241 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
