siRNA Details
Gene Symbol: Hist3h2bb-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206882.1 | NP_996765.1 | 1 | 167 | ||
GenBank mRNA | BC107301.1 | - | 1 | 166 | 40053183 | |
BC107302.1 | - | 1 | 167 | 40053184 |
Gene Symbol: Hist3h2ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030082.4 | NP_084358.1 | 1 | 112 | ||
GenBank mRNA | AK018765.1 | - | 1 | 113 | ||
BC051921.1 | - | 1 | 103 | 5684450 |

More Information
Other Species Targeted

Gene Ontology Categories:
