siRNA Details
Gene Symbol: Brd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289606.1 | NP_001276535.1 | 5 | 571 | ||
NM_001289607.1 | NP_001276536.1 | 5 | 571 | |||
NM_030147.3 | NP_084423.2 | 5 | 571 | |||
GenBank mRNA | AK032320.1 | - | 5 | 283 | ||
AK042446.1 | - | 5 | 297 | |||
AK132365.1 | - | 5 | 267 | |||
AK161689.1 | - | 5 | 284 | |||
BC021482.1 | - | 5 | 271 | |||
BC023837.1 | - | 5 | 287 | |||
BC025644.1 | - | 5 | 316 | 5323755 | ||
CD555707.1 | - | 5 | 571 |
