siRNA Details
Gene Symbol: Sfi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030207.2 | NP_084483.2 | 3, 4 | 399 | ||
GenBank mRNA | AK046135.1 | - | 3, 4 | 396 | ||
AK046373.1 | - | 3, 4 | 396 | |||
AK082866.1 | - | 3, 4 | 361 | |||
AK129159.1 | - | 3, 4 | 366 | |||
AK133558.1 | - | 3, 4 | 347 | |||
AK170090.1 | - | 3, 4 | 359 | |||
BC046305.1 | - | 3, 4 | 347 | 6493676 | ||
BC099677.1 | - | 3, 4 | 375 | |||
BC106124.1 | - | 425 |

More Information
Other Species Targeted

Gene Ontology Categories:
