siRNA Details
Gene Symbol: Fam122b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166365.2 | NP_001159837.2 | 2 | 983 | ||
NM_001166583.1 | NP_001160055.1 | 2 | 983 | |||
NM_030167.4 | NP_084443.1 | 2 | 983 | |||
GenBank mRNA | AB541013.1 | - | 2 | 151 | ||
AK019480.1 | - | 2 | 210 | |||
AK036417.1 | - | 2 | 980 | |||
AK044896.1 | - | 2 | 984 | |||
BC066167.1 | - | 2 | 198 | 30433483 |
