siRNA Details
Gene Symbol: Wrnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030215.3 | NP_084491.3 | 2 | 1134 | ||
GenBank mRNA | AB056151.1 | - | 2 | 983 | ||
AF208046.1 | - | 2 | 379 | |||
AK041886.1 | - | 2 | 1090 | |||
AK050368.1 | - | 2 | 1136 | |||
AK082078.1 | - | 2 | 1049 | |||
AK167570.1 | - | 2 | 1135 | |||
AK170542.1 | - | 2 | 1127 | |||
AK170846.1 | - | 2 | 1115 | |||
AK197211.1 | - | 1 | 126 | |||
AK210446.1 | - | 1 | 126 | |||
BC058744.1 | - | 2 | 524 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
