siRNA Details
Gene Symbol: Zfp687 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030074.2 | NP_084350.1 | 9 | 4416 | ||
GenBank mRNA | AK019851.2 | - | 9 | 4417 | ||
AK129360.1 | - | 3 | 4602 | |||
AK133173.1 | - | 6 | 5125 | |||
AK148239.1 | - | 8 | 4531 | |||
BC034652.1 | - | 3 | 1249 | |||
BC036979.1 | - | 4 | 1655 | |||
BC039950.1 | - | 4 | 1629 | |||
BC056943.1 | - | 7 | 4066 | 6831344 | ||
BC060179.1 | - | 7 | 4066 | 6831344 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008761341, XM_002729081, XM_006232934, XM_006232933, XM_006232935, and others.
Gene Ontology Categories:
