siRNA Details
Gene Symbol: Brd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289606.1 | NP_001276535.1 | 7 | 774 | ||
NM_001289607.1 | NP_001276536.1 | 7 | 774 | |||
NM_030147.3 | NP_084423.2 | 7 | 774 | |||
GenBank mRNA | AK032320.1 | - | 7 | 486 | ||
AK042446.1 | - | 7 | 500 | |||
AK132365.1 | - | 7 | 470 | |||
AK161689.1 | - | 7 | 487 | |||
BC021482.1 | - | 7 | 474 | |||
BC023837.1 | - | 7 | 490 | |||
BC025644.1 | - | 7 | 519 | 5323755 |

More Information
Other Species Targeted

Gene Ontology Categories:
