siRNA Details
Gene Symbol: Ddx19b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190786.1 | NP_001177715.1 | 12 | 6452 | ||
NM_001190800.1 | NP_001177729.1 | 12 | 6490 | |||
NM_172284.5 | NP_758488.1 | 12 | 6495 | |||
GenBank mRNA | AK014927.1 | - | 1 | 970 | ||
AK147499.1 | - | 12 | 6490 | |||
BC002300.1 | - | 1 | 1249 | |||
BC079644.1 | - | 1 | 1793 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
