siRNA Details
Gene Symbol: Psd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027626.1 | NP_081902.1 | 8 | 9069 | ||
NM_030263.5 | NP_084539.2 | 13 | 9411 | |||
NM_177698.4 | NP_808366.2 | 16 | 11175 | |||
GenBank mRNA | AK032036.1 | - | 1 | 2445 | ||
AK147606.1 | - | 8 | 9074 | |||
BC003498.1 | - | 1 | 923 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
