siRNA Details
Gene Symbol: Rcan2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286654.1 | NP_001273583.1 | 2, 3 | 216 | ||
NM_030598.2 | NP_085101.1 | 1, 2 | 346 | |||
GenBank mRNA | AB061524.1 | - | 1, 2 | 284 | ||
AF237791.1 | - | 1, 2 | 86 | |||
AF237887.1 | - | 1, 2 | 132 | |||
AK049006.1 | - | 1, 2 | 285 | |||
AK160865.1 | - | 1, 2 | 234 | |||
BC047153.1 | - | 1, 2 | 144 | 6491318 | ||
BC049096.1 | - | 1, 2 | 346 | 6416901 | ||
BC062141.1 | - | 2, 3 | 216 | 30290791 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
