siRNA Details
Gene Symbol: KALRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024660.4 | NP_001019831.2 | 36, 37 | 5721 | ||
NM_001322988.1 | NP_001309917.1 | 36, 37 | 5718 | |||
NM_001322993.1 | NP_001309922.1 | 3, 4 | 636 | |||
NM_001322994.1 | NP_001309923.1 | 3, 4 | 633 | |||
NM_001322995.1 | NP_001309924.1 | 3, 4 | 636 | |||
NM_001322996.1 | NP_001309925.1 | 3, 4 | 636 | |||
NM_001322997.1 | NP_001309926.1 | 3, 4 | 633 | |||
NM_007064.4 | NP_008995.2 | 3, 4 | 636 | |||
XM_006713810.3 | XP_006713873.1 | 36, 37 | 5966 | |||
XM_006713811.3 | XP_006713874.1 | 36, 37 | 5939 | |||
XM_006713812.3 | XP_006713875.1 | 36, 37 | 5927 | |||
XM_006713813.3 | XP_006713876.1 | 36, 37 | 5900 | |||
XM_011513279.2 | XP_011511581.1 | 36, 37 | 5966 | |||
XM_011513280.2 | XP_011511582.1 | 36, 37 | 5964 | |||
XM_011513281.2 | XP_011511583.1 | 36, 37 | 28588 | |||
XM_011513283.2 | XP_011511585.1 | 36, 37 | 5963 | |||
XM_017007429.1 | XP_016862918.1 | 36, 37 | 5966 | |||
XM_017007431.1 | XP_016862920.1 | 36, 37 | 5959 | |||
GenBank mRNA | AB011422.1 | - | 3, 4 | 590 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
