siRNA Details
Gene Symbol: Upf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122829.1 | NP_001116301.1 | 2 | 591 | ||
NM_030680.2 | NP_109605.2 | 2 | 591 | |||
GenBank mRNA | AF182947.1 | - | 3 | 580 | ||
AF322655.1 | - | 2 | 311 | |||
AK148196.1 | - | 3 | 568 | |||
AK205256.1 | - | 1 | 102 | |||
BC052149.1 | - | 2 | 529 | 6332288 | ||
BC056442.1 | - | 2 | 591 | 5702493 | ||
BQ771453.1 | - | 2 | 591 |

More Information
Other Species Targeted

Gene Ontology Categories:
