siRNA Details
Gene Symbol: Nptxr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030689.4 | NP_109614.2 | 3 | 1148 | ||
GenBank mRNA | AF316612.1 | - | 3 | 1130 | ||
AK147546.1 | - | 3 | 1205 | |||
AK147605.1 | - | 3 | 1156 | |||
AY394487.1 | - | 4 | 856 | |||
BC058962.1 | - | 3 | 1147 | 6826075 | ||
BC098199.1 | - | 3 | 1147 | 5705707 | ||
BC141096.1 | - | 3 | 1127 | 9055708 | ||
BF471950.1 | - | 3 | 451 |
Gene Symbol: Npcd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013360.2 | NP_001013378.2 | 6 | 781 | ||
NM_001013362.2 | NP_001013380.2 | 7 | 1072 | |||
GenBank mRNA | AY394488.1 | - | 6 | 686 | ||
AY394489.1 | - | 7 | 977 | |||
AY395907.1 | - | 9 | 1586 | |||
CD578976.1 | - | 2 | 387 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
