siRNA Details
Gene Symbol: Vmn1r56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030740.1 | NP_109665.1 | 1 | 410 | ||
GenBank mRNA | AF324868.1 | - | 1 | 410 |
Gene Symbol: Vmn1r55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166706.1 | NP_001160178.1 | 1 | 279 |
