siRNA Details
Gene Symbol: IL4R | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000418.3 | NP_000409.1 | 11 | 1230 | ||
NM_001257406.1 | NP_001244335.1 | 10 | 1097 | |||
NM_001257407.1 | NP_001244336.1 | 11 | 1341 | |||
NM_001257997.1 | NP_001244926.1 | 10 | 1092 | |||
XM_005255308.2 | XP_005255365.1 | 3 | 168 | |||
XM_006721043.1 | XP_006721106.1 | 2 | 267 | |||
XM_011545825.1 | XP_011544127.1 | 10 | 1135 | |||
XM_011545826.2 | XP_011544128.1 | 11 | 1186 | |||
XM_011545827.2 | XP_011544129.1 | 11 | 2425 | |||
XM_011545828.2 | XP_011544130.1 | 9 | 903 | |||
XM_011545829.1 | XP_011544131.1 | 10 | 1064 | |||
XM_011545830.2 | XP_011544132.1 | 10 | 2274 | |||
XM_011545831.1 | XP_011544133.1 | 9 | 983 | |||
XM_011545832.1 | XP_011544134.1 | 9 | 916 | |||
XM_011545833.2 | XP_011544135.1 | 8 | 834 | |||
XM_011545834.2 | XP_011544136.1 | 8 | 753 | |||
XM_017023211.1 | XP_016878700.1 | 9 | 1085 | |||
GenBank mRNA | AK223591.1 | - | 11 | 1148 | ||
AK303255.1 | - | 11 | 1316 | |||
AK314863.1 | - | 10 | 1082 | |||
BC136365.1 | - | 10 | 1129 | 9020352 | ||
BC151131.1 | - | 10 | 1129 | 9057031 | ||
X52425.1 | - | 11 | 1142 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
