siRNA Details
Gene Symbol: KALRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024660.4 | NP_001019831.2 | 60 | 10039 | ||
NM_001322993.1 | NP_001309922.1 | 27 | 4951 | |||
NM_007064.4 | NP_008995.2 | 27 | 4954 | |||
XM_006713810.3 | XP_006713873.1 | 60 | 10284 | |||
XM_006713811.3 | XP_006713874.1 | 60 | 10257 | |||
XM_006713812.3 | XP_006713875.1 | 60 | 10245 | |||
XM_006713813.3 | XP_006713876.1 | 60 | 10218 | |||
XM_006713814.3 | XP_006713877.1 | 59 | 10188 | |||
XM_011513279.2 | XP_011511581.1 | 60 | 10281 | |||
XM_011513280.2 | XP_011511582.1 | 60 | 10282 | |||
XM_011513281.2 | XP_011511583.1 | 60 | 32906 | |||
XM_017007429.1 | XP_016862918.1 | 59 | 10224 | |||
XM_017007430.1 | XP_016862919.1 | 59 | 10185 | |||
GenBank mRNA | AB011422.1 | - | 27 | 4909 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
