siRNA Details
Gene Symbol: Acox3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030721.2 | NP_109646.2 | 4 | 445 | ||
GenBank mRNA | AJ278430.1 | - | 4 | 478 | ||
AK028818.1 | - | 4 | 866 | |||
AK053960.1 | - | 4 | 442 | |||
AK128922.1 | - | 4 | 405 | |||
AK154112.1 | - | 3 | 536 | |||
AK171598.1 | - | 428 | ||||
AK194193.1 | - | 1 | 83 | |||
BC044725.2 | - | 4 | 465 | 5374281 | ||
BC055019.1 | - | 4 | 445 | 4162223 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
