siRNA Details
Gene Symbol: Vmn1r172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030735.1 | NP_109660.1 | 2 | 649 | ||
GenBank mRNA | AF324873.1 | - | 2 | 649 | ||
BC139316.1 | - | 2 | 644 | 8862338 | ||
BC139317.1 | - | 2 | 644 | 8862339 | ||
BC145601.1 | - | 2 | 644 | 9054167 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
