siRNA Details
Gene Symbol: Mynn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289622.1 | NP_001276551.1 | 5, 6 | 1917 | ||
NM_030557.3 | NP_085034.2 | 5, 6 | 2047 | |||
GenBank mRNA | AF349561.1 | - | 4, 5 | 1439 | ||
AK019238.2 | - | 5, 6 | 1491 | |||
AK029657.1 | - | 5, 6 | 1338 | |||
AK136107.1 | - | 5, 6 | 1496 | |||
AK163261.1 | - | 5, 6 | 1361 | |||
AK163264.1 | - | 5, 6 | 1361 | |||
BC007477.1 | - | 5, 6 | 1360 | |||
BC065084.1 | - | 5, 6 | 2047 | 6848601 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006232234, XM_006232232.
Gene Ontology Categories:
