siRNA Details
Gene Symbol: Senp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163571.1 | NP_001157043.1 | 11 | 2218 | ||
NM_030702.4 | NP_109627.3 | 11 | 2430 | |||
GenBank mRNA | AY008764.1 | - | 11 | 2162 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
