siRNA Details
Gene Symbol: Cxcr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030712.4 | NP_109637.3 | 2 | 428 | ||
GenBank mRNA | AF301018.1 | - | 2 | 431 | ||
AF305709.1 | - | 2 | 431 | |||
AK052901.1 | - | 2 | 426 | |||
AK137703.1 | - | 2 | 407 | |||
AK153914.1 | - | 2 | 427 | |||
JC209414.1 | - | 2 | 428 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
