siRNA Details
Gene Symbol: Vmn1r172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030735.1 | NP_109660.1 | 2 | 1165 | ||
GenBank mRNA | AF324873.1 | - | 2 | 1165 | ||
BC139316.1 | - | 2 | 1160 | 8862338 | ||
BC139317.1 | - | 2 | 1160 | 8862339 | ||
BC145601.1 | - | 2 | 1160 | 9054167 |
Gene Symbol: Vmn1r173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166718.1 | NP_001160190.1 | 1 | 912 |
Gene Symbol: Vmn1r174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207548.2 | NP_997431.2 | 1 | 912 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
